siRNA Details
Gene Symbol: PDGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002607.5 | NP_002598.4 | 5 | 1312 | ||
NM_033023.4 | NP_148983.1 | 5 | 1312 | |||
XM_011515415.1 | XP_011513717.1 | 6 | 804 | |||
XM_011515416.1 | XP_011513718.1 | 6 | 808 | |||
XM_011515417.1 | XP_011513719.1 | 6 | 804 | |||
XM_011515418.1 | XP_011513720.1 | 6 | 808 | |||
XM_011515419.2 | XP_011513721.1 | 5 | 512 | |||
XM_017012289.1 | XP_016867778.1 | 5 | 545 | |||
GenBank mRNA | A09204.1 | - | 5 | 505 | ||
AB451309.1 | - | 5 | 469 | |||
AB451439.1 | - | 5 | 469 | |||
AK292217.1 | - | 5 | 683 | |||
AU118732.1 | - | 5 | 640 | |||
BC109246.1 | - | 3 | 1332 | |||
BE336646.1 | - | 4 | 426 | |||
X03795.1 | - | 5 | 856 | |||
X06374.1 | - | 5 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
