siRNA Details
Gene Symbol: CARMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126481.1 | - | 5 | 2443 | ||
AL137734.1 | - | 1 | 2782 | |||
CR933654.1 | - | 2 | 5407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
