siRNA Details
Gene Symbol: ZNRD1ASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF032109.2 | - | 3 | 3307 | ||
AF032111.1 | - | 1 | 380 | |||
BX648746.1 | - | 3 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
