siRNA Details
Gene Symbol: SULT1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321770.1 | NP_001308699.1 | 1 | 504 | ||
NM_006588.3 | NP_006579.2 | 1 | 504 | |||
NR_135776.1 | 1 | 504 | ||||
NR_135779.1 | 1 | 504 | ||||
GenBank mRNA | AK297851.1 | - | 1 | 282 | ||
AK298755.1 | - | 1 | 445 | |||
BC058861.1 | - | 1 | 504 | |||
BC125043.1 | - | 1 | 230 | 40115974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
