siRNA Details
Gene Symbol: LOC100506271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110515.1 | 1 | 911 | |||
GenBank mRNA | AK097787.1 | - | 1 | 713 | ||
BC043218.2 | - | 1 | 912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
