siRNA Details
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 13 | 2733 | ||
XM_011517056.2 | XP_011515358.1 | 14 | 3539 | |||
XM_011517058.2 | XP_011515360.1 | 13 | 3394 | |||
XM_011517059.1 | XP_011515361.1 | 13 | 2661 | |||
XM_011517060.2 | XP_011515362.1 | 14 | 3521 | |||
XM_011517061.2 | XP_011515363.1 | 14 | 2878 | |||
XM_011517062.1 | XP_011515364.1 | 13 | 2733 | |||
XM_011517063.2 | XP_011515365.1 | 13 | 3376 | |||
XM_011517064.2 | XP_011515366.1 | 14 | 3538 | |||
XM_011517065.1 | XP_011515367.1 | 13 | 2797 | |||
XM_011517068.2 | XP_011515370.1 | 13 | 2626 | |||
XM_011517069.2 | XP_011515371.1 | 8 | 2720 | |||
XM_011517070.2 | XP_011515372.1 | 14 | 3543 | |||
XM_011517072.2 | XP_011515374.1 | 7 | 1978 | |||
XM_011517073.2 | XP_011515375.1 | 7 | 1982 | |||
XM_011517074.1 | XP_011515376.1 | 6 | 1848 | |||
XM_017013470.1 | XP_016868959.1 | 13 | 3345 | |||
XM_017013471.1 | XP_016868960.1 | 14 | 3482 | |||
GenBank mRNA | AB196635.1 | - | 13 | 2734 | ||
AF131744.1 | - | 1 | 492 | |||
AK091433.1 | - | 13 | 2733 | |||
BC037288.1 | - | 14 | 2768 | 5266568 | ||
BC106912.1 | - | 1 | 299 | |||
BC106913.1 | - | 1 | 299 | |||
CR457147.1 | - | 1 | 162 | |||
CR936831.1 | - | 7 | 1926 |
