siRNA Details
Gene Symbol: ERVK13-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040023.1 | 3 | 5283 | |||
GenBank mRNA | AK000400.1 | - | 1 | 953 | ||
AK091794.1 | - | 3 | 3075 | |||
AK095440.1 | - | 1 | 2880 | |||
AK097909.1 | - | 1 | 1338 | |||
AK131509.1 | - | 1 | 3909 | |||
AK292678.1 | - | 1 | 1856 | |||
AX747190.1 | - | 3 | 3075 | |||
BC043252.1 | - | 3 | 5285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
