siRNA Details
Gene Symbol: ERVK13-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040023.1 | 3 | 4288 | |||
GenBank mRNA | AK091794.1 | - | 3 | 2080 | ||
AK095440.1 | - | 1 | 1885 | |||
AK097014.1 | - | 2 | 1820 | |||
AK097909.1 | - | 1 | 343 | |||
AK131509.1 | - | 1 | 2914 | |||
AK292678.1 | - | 1 | 861 | |||
AX747190.1 | - | 3 | 2080 | |||
BC043252.1 | - | 3 | 4290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
