siRNA Details
Gene Symbol: DKFZP434L187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026771.1 | 3 | 4140 | |||
GenBank mRNA | AL117445.1 | - | 2 | 4029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
