siRNA Details
Gene Symbol: ANTXRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278688.2 | NP_001265617.1 | 2 | 561 | ||
XM_006717684.3 | XP_006717747.1 | 2 | 561 | |||
XM_011539430.2 | XP_011537732.1 | 2 | 561 | |||
XM_011539441.1 | XP_011537743.1 | 2 | 561 | |||
XM_011539442.1 | XP_011537744.1 | 2 | 561 | |||
GenBank mRNA | AK057316.1 | - | 3 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
