siRNA Details
Gene Symbol: MACROD2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104193.2 | 2, 3 | 203 | |||
GenBank mRNA | CR747489.1 | - | 2, 3 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
