siRNA Details
Gene Symbol: LOC646029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130930.1 | 3 | 407 | |||
GenBank mRNA | AK096213.1 | - | 2 | 351 | ||
CD557269.1 | - | 3 | 211 | |||
DA680640.1 | - | 3 | 218 | |||
DB028980.1 | - | 3 | 340 | |||
DB052548.1 | - | 3 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
