siRNA Details
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014979.2 | NP_001014979.2 | 6 | 1115 | ||
XM_011545976.1 | XP_011544278.1 | 6 | 1148 | |||
XM_011545978.1 | XP_011544280.1 | 5 | 1063 | |||
XM_017023852.1 | XP_016879341.1 | 5 | 1343 | |||
XM_017023853.1 | XP_016879342.1 | 4 | 1280 | |||
XM_017023854.1 | XP_016879343.1 | 5 | 1343 | |||
XM_017023855.1 | XP_016879344.1 | 6 | 1247 | |||
XM_017023856.1 | XP_016879345.1 | 5 | 864 | |||
XM_017023857.1 | XP_016879346.1 | 5 | 1424 | |||
XM_017023858.1 | XP_016879347.1 | 5 | 1343 | |||
XM_017023859.1 | XP_016879348.1 | 6 | 1219 | |||
XM_017023860.1 | XP_016879349.1 | 2 | 518 | |||
XM_017023861.1 | XP_016879350.1 | 6 | 1219 | |||
XM_017023862.1 | XP_016879351.1 | 4 | 1661 | |||
GenBank mRNA | AL122109.2 | - | 4 | 1621 | ||
BC042548.1 | - | 4 | 1420 | |||
BC092491.1 | - | 5 | 1407 | 4837696 | ||
BC128128.1 | - | 5 | 771 | |||
BC128129.1 | - | 6 | 619 | 40112368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
