siRNA Details
Gene Symbol: LINC00575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024087.1 | 4 | 759 | |||
NR_024088.1 | 4 | 697 | ||||
GenBank mRNA | AY316301.1 | - | 4 | 759 | ||
AY316302.1 | - | 4 | 697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
