siRNA Details
Gene Symbol: C8orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103552.1 | 1 | 1368 | |||
GenBank mRNA | AK055534.1 | - | 1 | 1370 | ||
BC132945.1 | - | 1 | 1235 | 40146967 | ||
BC136882.1 | - | 1 | 1239 | 9020873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
