siRNA Details
Gene Symbol: KLHDC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014997.3 | NP_055812.1 | 10 | 6207 | ||
XM_005250233.4 | XP_005250290.1 | 9 | 6096 | |||
GenBank mRNA | AF277177.1 | - | 1 | 603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
