siRNA Details
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 9497 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 8563 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 9364 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 9051 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 9626 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 9890 | |||
NM_003879.5 | NP_003870.4 | 10 | 9469 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 10299 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 12316 | |||
GenBank mRNA | AK124664.1 | - | 1 | 1669 | ||
AK125394.1 | - | 1 | 2196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
