siRNA Details
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024127.1 | 4 | 516 | |||
GenBank mRNA | AF277181.1 | - | 4 | 304 | ||
AK092096.1 | - | 2 | 1361 | |||
AY277594.1 | - | 2 | 84 | |||
BC022205.1 | - | 4 | 1145 | |||
BC034611.1 | - | 2 | 1367 | |||
BC045792.1 | - | 4 | 551 | |||
BC064589.1 | - | 3 | 1082 | |||
BC068048.1 | - | 4 | 516 | |||
BC107730.1 | - | 1 | 908 | |||
KJ782215.1 | - | 5 | 1645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
