siRNA Details
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024127.1 | 6 | 656 | |||
GenBank mRNA | AF277181.1 | - | 6 | 444 | ||
AK092096.1 | - | 3 | 1825 | |||
AY277594.1 | - | 4 | 224 | |||
BC022205.1 | - | 6 | 1285 | |||
BC034611.1 | - | 4 | 1507 | |||
BC045792.1 | - | 5 | 1015 | |||
BC064589.1 | - | 4 | 1546 | |||
BC068048.1 | - | 6 | 656 | |||
BC107730.1 | - | 3 | 1048 | |||
KJ782215.1 | - | 7 | 1785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
