siRNA Details
Gene Symbol: APOL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030641.3 | NP_085144.1 | 3 | 9654 | ||
XM_011530392.2 | XP_011528694.1 | 4 | 9831 | |||
GenBank mRNA | AK097266.1 | - | 1 | 1766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
