siRNA Details
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015630.3 | NP_056445.3 | 12, 13 | 2052 | ||
XM_011510941.2 | XP_011509243.1 | 12, 13 | 2257 | |||
XM_011510943.2 | XP_011509245.1 | 11, 12 | 1936 | |||
GenBank mRNA | AF286904.1 | - | 12, 13 | 2044 | ||
AK001433.1 | - | 10, 11 | 1643 | |||
AK002010.1 | - | 3, 4 | 499 | |||
AK125789.1 | - | 13, 14 | 2121 | |||
AL117558.1 | - | 1, 2 | 46 | |||
BC093818.1 | - | 12, 13 | 1964 | 7939663 | ||
BC093820.1 | - | 12, 13 | 1964 | 7939665 |
