siRNA Details
Gene Symbol: TTC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242672.1 | NP_001229601.1 | 5 | 981 | ||
XM_016999990.1 | XP_016855479.1 | 5 | 1053 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
