siRNA Details
Gene Symbol: TMEM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014698.2 | NP_055513.2 | 8 | 1174 | ||
XM_011544328.2 | XP_011542630.1 | 8 | 1026 | |||
XM_011544329.2 | XP_011542631.1 | 8 | 2293 | |||
XM_011544330.2 | XP_011542632.1 | 7 | 983 | |||
XM_011544331.2 | XP_011542633.1 | 8 | 857 | |||
XM_017002916.1 | XP_016858405.1 | 8 | 828 | |||
GenBank mRNA | AB018335.1 | - | 8 | 802 | ||
AK022502.1 | - | 8 | 823 | |||
AK222934.1 | - | 8 | 804 | |||
AK222944.1 | - | 8 | 804 | |||
AK309210.1 | - | 6 | 751 | |||
BC030245.1 | - | 8 | 807 | 5216414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
