siRNA Details
Gene Symbol: TMEM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014698.2 | NP_055513.2 | 25 | 3682 | ||
XM_006711841.3 | XP_006711904.1 | 23 | 3175 | |||
XM_011544328.2 | XP_011542630.1 | 25 | 3534 | |||
XM_011544329.2 | XP_011542631.1 | 25 | 4801 | |||
XM_011544330.2 | XP_011542632.1 | 24 | 3491 | |||
XM_011544331.2 | XP_011542633.1 | 24 | 3278 | |||
XM_011544332.2 | XP_011542634.1 | 24 | 3313 | |||
XM_017002916.1 | XP_016858405.1 | 24 | 3765 | |||
GenBank mRNA | AB018335.1 | - | 25 | 3310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
