siRNA Details
Gene Symbol: ZNF252P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023392.1 | 5 | 481 | |||
GenBank mRNA | AK294723.1 | - | 3 | 224 | ||
AK296398.1 | - | 5 | 477 | |||
BC019922.1 | - | 1 | 15 | |||
BI254488.1 | - | 1 | 15 | |||
DA489661.1 | - | 5 | 481 | |||
DB233439.1 | - | 5 | 478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
