siRNA Details
Gene Symbol: SETD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015048.1 | NP_055863.1 | 2 | 241 | ||
XM_005253858.4 | XP_005253915.1 | 3 | 400 | |||
XM_006719296.3 | XP_006719359.1 | 3 | 316 | |||
GenBank mRNA | JF813787.1 | - | 2 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
