siRNA Details
Gene Symbol: LINC01529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104176.1 | 3 | 333 | |||
GenBank mRNA | AK055260.1 | - | 3 | 309 | ||
DA464500.1 | - | 3 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
