siRNA Details
Gene Symbol: TMEM30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017970.2 | NP_001017970.1 | 1 | 2068 | ||
GenBank mRNA | AK091169.1 | - | 1 | 1497 | ||
DA431345.1 | - | 1 | 75 | |||
DA666826.1 | - | 1 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
