siRNA Details
Gene Symbol: PCNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032223.3 | NP_115599.2 | 27 | 4394 | ||
XM_005273987.4 | XP_005274044.1 | 27 | 6314 | |||
XM_006718549.3 | XP_006718612.1 | 27 | 6299 | |||
XM_006718551.3 | XP_006718614.1 | 27 | 6317 | |||
XM_011545024.2 | XP_011543326.1 | 28 | 5490 | |||
XM_011545025.2 | XP_011543327.1 | 27 | 6317 | |||
XM_011545026.2 | XP_011543328.1 | 21 | 3157 | |||
XM_011545027.1 | XP_011543329.1 | 19 | 2370 | |||
GenBank mRNA | BX640978.1 | - | 7 | 1066 | ||
CA489689.1 | - | 4 | 583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
