siRNA Details
Gene Symbol: PCNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032223.3 | NP_115599.2 | 35 | 6134 | ||
XM_005273987.4 | XP_005274044.1 | 35 | 8054 | |||
XM_006718549.3 | XP_006718612.1 | 35 | 8039 | |||
XM_011545024.2 | XP_011543326.1 | 36 | 7230 | |||
XM_011545026.2 | XP_011543328.1 | 29 | 4897 | |||
XM_011545027.1 | XP_011543329.1 | 27 | 4110 | |||
GenBank mRNA | BX640978.1 | - | 15 | 2806 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
