siRNA Details
Gene Symbol: RTEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283009.1 | NP_001269938.1 | 29, 30 | 3686 | ||
NM_001283010.1 | NP_001269939.1 | 28, 29 | 3407 | |||
NM_016434.3 | NP_057518.1 | 29, 30 | 3686 | |||
NM_032957.4 | NP_116575.3 | 29, 30 | 3758 | |||
GenBank mRNA | AF217795.1 | - | 29, 30 | 3686 | ||
AK000485.1 | - | 5, 6 | 889 | |||
AK299332.1 | - | 29, 30 | 3181 | |||
AK302508.1 | - | 29, 30 | 3277 | |||
AK304798.1 | - | 28, 29 | 2903 | |||
AL080127.1 | - | 6, 7 | 739 | |||
JX119186.1 | - | 28, 29 | 2859 |
Gene Symbol: RTEL1-TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037882.1 | 29, 30 | 3686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
