siRNA Details
Gene Symbol: ZNF778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023016.1 | XP_016878505.1 | 8 | 7498 | ||
XM_017023017.1 | XP_016878506.1 | 8 | 7397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
