siRNA Details
Gene Symbol: MEIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080836.2 | NP_001074305.1 | 3 | 336 | ||
GenBank mRNA | BG189536.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
