siRNA Details
Gene Symbol: SIGLEC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002825.2 | 7 | 1432 | |||
GenBank mRNA | AK301031.1 | - | 7 | 1436 | ||
BC039008.1 | - | 7 | 1363 | |||
EU330985.1 | - | 7 | 1345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
