siRNA Details
Gene Symbol: LOC254028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122043.1 | 5 | 1234 | |||
GenBank mRNA | AK093210.1 | - | 4 | 1179 | ||
BX247965.1 | - | 3 | 740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
