siRNA Details
Gene Symbol: CMYA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153610.4 | NP_705838.3 | 2 | 3353 | ||
XM_017009212.1 | XP_016864701.1 | 2 | 3221 | |||
GenBank mRNA | AK092699.1 | - | 1 | 84 | ||
AL832347.1 | - | 2 | 3155 | |||
AL832376.1 | - | 2 | 3312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
