siRNA Details
Gene Symbol: CYP4F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007253.3 | NP_009184.1 | 4 | 432 | ||
XM_017026232.1 | XP_016881721.1 | 3 | 369 | |||
GenBank mRNA | AF133298.1 | - | 3 | 375 | ||
AK096809.1 | - | 2 | 2117 | |||
AK300530.1 | - | 4 | 432 | |||
AK314685.1 | - | 4 | 430 |
