siRNA Details
Gene Symbol: ZDBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285549.1 | NP_001272478.1 | 7 | 2132 | ||
NM_020923.2 | NP_065974.1 | 5 | 2239 | |||
XM_005246711.3 | XP_005246768.1 | 6 | 2239 | |||
XM_005246712.4 | XP_005246769.1 | 6 | 2062 | |||
XM_005246713.3 | XP_005246770.1 | 6 | 2056 | |||
XM_006712656.3 | XP_006712719.1 | 6 | 2095 | |||
XM_011511532.2 | XP_011509834.1 | 6 | 2272 | |||
XM_011511533.2 | XP_011509835.1 | 6 | 2089 | |||
XM_011511534.2 | XP_011509836.1 | 5 | 2012 | |||
XM_011511535.2 | XP_011509837.1 | 5 | 2018 | |||
XM_017004573.1 | XP_016860062.1 | 7 | 2345 | |||
XM_017004574.1 | XP_016860063.1 | 7 | 2271 | |||
XM_017004575.1 | XP_016860064.1 | 6 | 2240 | |||
XM_017004576.1 | XP_016860065.1 | 1 | 412 | |||
GenBank mRNA | AB046791.2 | - | 1 | 107 | ||
AB775780.1 | - | 7 | 2133 | |||
AB775781.1 | - | 5 | 2239 | |||
AK127271.1 | - | 5 | 2084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
