siRNA Details
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317916.1 | NP_001304845.1 | 4, 5 | 555 | ||
NM_033160.6 | NP_149350.3 | 4, 5 | 398 | |||
NR_134255.1 | 3, 4 | 443 | ||||
XM_005272515.4 | XP_005272572.1 | 4, 5 | 505 | |||
XM_011545679.2 | XP_011543981.1 | 4, 5 | 329 | |||
XM_017014614.1 | XP_016870103.1 | 3, 4 | 2691 | |||
GenBank mRNA | DA120232.1 | - | 4, 5 | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
