siRNA Details
Gene Symbol: CCDC154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143980.1 | NP_001137452.1 | 11, 12 | 1465 | ||
GenBank mRNA | JN935901.1 | - | 10, 11 | 1298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
