siRNA Details
Gene Symbol: ZNF527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032453.1 | NP_115829.1 | 5 | 1478 | ||
XM_005259328.4 | XP_005259385.1 | 5 | 1570 | |||
XM_005259329.3 | XP_005259386.1 | 4 | 1484 | |||
XM_017027380.1 | XP_016882869.1 | 3 | 1459 | |||
XM_017027381.1 | XP_016882870.1 | 4 | 1470 | |||
GenBank mRNA | AB058732.1 | - | 2 | 1211 | ||
AK091585.1 | - | 4 | 1373 | |||
AK301135.1 | - | 5 | 1474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
