siRNA Details
Gene Symbol: MUC5AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304359.1 | NP_001291288.1 | 741 | |||
GenBank mRNA | AF015521.1 | - | 7 | 741 | ||
AF043909.1 | - | 7 | 730 | |||
AJ298317.1 | - | 7 | 732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
