siRNA Details
Gene Symbol: LOC100129940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292023.1 | NP_001278952.1 | 1 | 267 | ||
NM_001292024.1 | NP_001278953.1 | 1 | 267 | |||
GenBank mRNA | BI762425.1 | - | 1 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
