siRNA Details
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 32 | 4709 | ||
NM_173636.4 | NP_775907.4 | 32 | 4694 | |||
XM_005258809.2 | XP_005258866.1 | 31 | 4598 | |||
XM_011526837.1 | XP_011525139.1 | 32 | 4694 | |||
XM_011526838.1 | XP_011525140.1 | 30 | 4460 | |||
XM_011526839.1 | XP_011525141.1 | 30 | 4358 | |||
XM_011526840.2 | XP_011525142.1 | 25 | 3704 | |||
XM_011526841.2 | XP_011525143.1 | 23 | 3325 | |||
XM_011526842.1 | XP_011525144.1 | 22 | 3157 | |||
XM_011526843.1 | XP_011525145.1 | 15 | 2484 | |||
XM_011526844.2 | XP_011525146.1 | 15 | 2481 | |||
GenBank mRNA | AL117438.1 | - | 6 | 1399 | ||
AL133651.1 | - | 11 | 2006 | |||
BC017261.2 | - | 14 | 2403 | |||
BC058939.1 | - | 32 | 4653 | |||
BX647726.1 | - | 30 | 5940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
