siRNA Details
Gene Symbol: PLAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182832.2 | NP_878252.2 | 1 | 4740 | ||
GenBank mRNA | AF269287.1 | - | 1 | 989 | ||
AK027868.1 | - | 1 | 2402 | |||
L13197.1 | - | 1 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
