siRNA Details
Gene Symbol: SLC22A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039752.3 | NP_001034841.3 | 3, 4 | 866 | ||
NR_134874.1 | 4, 5 | 621 | ||||
XM_011545016.2 | XP_011543318.1 | 5, 6 | 1871 | |||
XM_017017699.1 | XP_016873188.1 | 4, 5 | 668 | |||
XM_017017701.1 | XP_016873190.1 | 5, 6 | 1871 | |||
XM_017017702.1 | XP_016873191.1 | 5, 6 | 1871 | |||
XM_017017703.1 | XP_016873192.1 | 2, 3 | 212 | |||
GenBank mRNA | AB075876.1 | - | 4, 5 | 621 | ||
AK296299.1 | - | 5, 6 | 573 | |||
AK298430.1 | - | 4, 5 | 668 | |||
DR003989.1 | - | 3, 4 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
