siRNA Details
Gene Symbol: SLC22A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039752.3 | NP_001034841.3 | 9 | 1700 | ||
NR_134874.1 | 10 | 1316 | ||||
XM_011545016.2 | XP_011543318.1 | 11 | 2705 | |||
XM_017017699.1 | XP_016873188.1 | 10 | 1502 | |||
XM_017017700.1 | XP_016873189.1 | 9 | 1938 | |||
XM_017017701.1 | XP_016873190.1 | 11 | 2566 | |||
XM_017017703.1 | XP_016873192.1 | 8 | 1046 | |||
XM_017017704.1 | XP_016873193.1 | 9 | 1798 | |||
GenBank mRNA | AB075876.1 | - | 10 | 1316 | ||
AK298430.1 | - | 10 | 1363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
