siRNA Details
Gene Symbol: ZNF879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136116.1 | NP_001129588.1 | 5 | 2022 | ||
XM_005265908.3 | XP_005265965.1 | 5 | 2716 | |||
XM_011534550.2 | XP_011532852.1 | 5 | 2051 | |||
XM_011534551.2 | XP_011532853.1 | 5 | 2167 | |||
GenBank mRNA | AK300504.1 | - | 5 | 1954 | ||
BX648737.1 | - | 5 | 1992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
