siRNA Details
Gene Symbol: ZNF879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136116.1 | NP_001129588.1 | 5 | 2247 | ||
XM_005265908.3 | XP_005265965.1 | 5 | 2941 | |||
XM_011534550.2 | XP_011532852.1 | 5 | 2276 | |||
XM_011534551.2 | XP_011532853.1 | 5 | 2392 | |||
GenBank mRNA | BX114675.1 | - | 1 | 163 | ||
BX648737.1 | - | 5 | 2217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
