siRNA Details
Gene Symbol: ZNF772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024596.2 | NP_001019767.1 | 5 | 871 | ||
NM_001144068.1 | NP_001137540.1 | 4 | 748 | |||
XM_005258943.4 | XP_005259000.1 | 3 | 4001 | |||
XM_005258944.3 | XP_005259001.1 | 2 | 3962 | |||
XM_011526980.2 | XP_011525282.1 | 5 | 4505 | |||
XM_017026813.1 | XP_016882302.1 | 4 | 4382 | |||
GenBank mRNA | AK293825.1 | - | 3 | 671 | ||
AK294941.1 | - | 3 | 576 | |||
AK301562.1 | - | 2 | 550 | |||
AK302574.1 | - | 4 | 710 | |||
BX647068.1 | - | 5 | 871 | |||
CR749225.1 | - | 2 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
